accessibility menu, dialog, popup

UserName

CAS RN 67-72-1

IUPAC Name: hexachloroethane
Synonyms: hexachloroethane hexachloraethan C2cl6 carbon hexachloride
Molecular Weight (g/mol): 236.72
Molecular Formula: C2Cl6
InChi Key: VHHHONWQHHHLTI-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(Cl)(Cl)Cl
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)