missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroethane 99.0+%, TCI America™

Catalog No. H0060500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0060500G 500 g
H006025G 25 g
H0060100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0060500G Supplier TCI America Supplier No. H0060500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-72-1
Molecular Formula C2Cl6
Molecular Weight (g/mol) 236.72
MDL Number MFCD00000799
InChI Key VHHHONWQHHHLTI-UHFFFAOYSA-N
Synonym hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin
PubChem CID 6214
ChEBI CHEBI:39227
IUPAC Name hexachloroethane
SMILES ClC(Cl)(Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 185°C
Color White
Quantity 500 g
UN Number 3077
Formula Weight 236.72
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexachloroethane
EINECSNumber (2)-0057
RTECSNumber KI4025000
TSCA Yes