accessibility menu, dialog, popup

UserName

CAS RN 66872-75-1

IUPAC Name: calcium bis(3-methyl-2-oxopentanoate)
Synonyms: calcium bis(3-methyl-2-oxopentanoate)
Molecular Weight (g/mol): 298.35
Molecular Formula: C12H18CaO6
InChi Key: PTFSVYLXDCGPFY-UHFFFAOYNA-L
SMILES: [Ca++].CCC(C)C(=O)C([O-])=O.CCC(C)C(=O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results