missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-Methyl-2-oxovalerate Hydrate 98.0+%, TCI America™

Catalog No. K00225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00225G 5 g
K002225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00225G Supplier TCI America Supplier No. K00225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66872-75-1
Molecular Formula C12H18CaO6
Molecular Weight (g/mol) 298.348
MDL Number MFCD00002583
InChI Key PTFSVYLXDCGPFY-UHFFFAOYSA-L
Synonym 3-Methyl-2-oxovaleric Acid Calcium Salt
PubChem CID 6099313
IUPAC Name calcium;3-methyl-2-oxopentanoate
SMILES CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-].[Ca+2]

Specifications

Color White
Quantity 5 g
Formula Weight 298.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium 3-Methyl-2-oxovalerate Hydrate