accessibility menu, dialog, popup

UserName

CAS RN 66684-58-0

IUPAC Name: 1,2,3-trifluoro-5-nitrobenzene
Synonyms: 1,2,3-trifluoro-5-nitrobenzene
Molecular Weight (g/mol): 177.08
Molecular Formula: C6H2F3NO2
InChi Key: PTTUMBGORBMNBN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)