missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluoro-5-nitrobenzene, 99%

Catalog No. AAL2030214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2030214 25 g
AAL2030206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2030214 Supplier Thermo Scientific Chemicals Supplier No. L2030214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 66684-58-0 | C6H2F3NO2 | 177.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66684-58-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00456803
InChI Key PTTUMBGORBMNBN-UHFFFAOYSA-N
Synonym 3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
PubChem CID 2782793
IUPAC Name 1,2,3-trifluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(F)C(F)=C1

Specifications

Density 1.52
Flash Point 75°C (167°F)
Refractive Index 1.486
Quantity 25 g
UN Number UN2810
Beilstein 2455018
Formula Weight 177.08
Percent Purity 99%
Chemical Name or Material 1,2,3-Trifluoro-5-nitrobenzene
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 266-447-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only