accessibility menu, dialog, popup

UserName

CAS RN 66640-86-6

IUPAC Name: 5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanehydrazide
Synonyms: 5-[(3aS,4S,6aR)-2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl]pentanehydrazide
Molecular Weight (g/mol): 258.34
Molecular Formula: C10H18N4O2S
InChi Key: KOZWHQPRAOJMBN-ZKWXMUAHSA-N
SMILES: NNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results