missing translation for 'onlineSavingsMsg'
Learn More

Biotin Hydrazide 98.0+%, TCI America™

Catalog No. B243125MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B243125MG 25 mg
B2431100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B243125MG Supplier TCI America Supplier No. B243125MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66640-86-6
Molecular Formula C10H18N4O2S
Molecular Weight (g/mol) 258.34
MDL Number MFCD00078532
InChI Key KOZWHQPRAOJMBN-ZKWXMUAHSA-N
Synonym biotin hydrazide, +-biotin hydrazide, 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanehydrazide, biotin-hydrazide, 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanehydrazi de, 5-3as,4s,6ar-2-oxo-1,3,3a,4,6,6a-hexahydrothieno 3,4-d imidazol-4-yl pentanehydrazide, biotin hydrazone, 5-3as,4s,6ar-2-oxo-hexahydrothieno 3,4-d imidazol-4-yl pentanehydrazide, biotin-nhnh2
PubChem CID 83872
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanehydrazide
SMILES C1C2C(C(S1)CCCCC(=O)NN)NC(=O)N2

Specifications

Melting Point 233°C
Color White
Quantity 25 mg
Formula Weight 258.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin Hydrazide
TSCA No
Recommended Storage Refrigerator