accessibility menu, dialog, popup

UserName

CAS RN 664302-70-9

IUPAC Name: 1-(isoquinolin-1-yl)naphthalen-2-amine
Synonyms: 1-(isoquinolin-1-yl)naphthalen-2-amine
Molecular Weight (g/mol): 270.34
Molecular Formula: C19H14N2
InChi Key: OYLNFFGFQUEPML-UHFFFAOYSA-N
SMILES: NC1=C(C2=CC=CC=C2C=C1)C1=C2C=CC=CC2=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results