missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Amino-1-naphthyl)isoquinoline 98.0+%, TCI America™

Catalog No. A22211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22211G Supplier TCI America Supplier No. A22211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 664302-70-9
Molecular Formula C19H14N2
Molecular Weight (g/mol) 270.335
InChI Key OYLNFFGFQUEPML-UHFFFAOYSA-N
Synonym 1-(Isoquinolinyl)-2-naphthylamine
PubChem CID 24938303
IUPAC Name 1-isoquinolin-1-ylnaphthalen-2-amine
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=NC=CC4=CC=CC=C43)N

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 270.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Amino-1-naphthyl)isoquinoline