accessibility menu, dialog, popup

UserName

CAS RN 66416-72-6

IUPAC Name: 4-bromo-2-iodoaniline
Synonyms: 4-bromo-2-iodoaniline
Molecular Weight (g/mol): 297.92
Molecular Formula: C6H5BrIN
InChi Key: HHTYEQWCHQEJNV-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)