accessibility menu, dialog, popup

UserName

OOOOOHOH

CAS RN 66-76-2

IUPAC Name: 4-hydroxy-3-[(4-hydroxy-2-oxo-2H-chromen-3-yl)methyl]-2H-chromen-2-one
Synonyms: dicoumarol dicoumarolum dicoumarol (inn)
Molecular Weight (g/mol): 336.3
Molecular Formula: C19H12O6
InChi Key: DOBMPNYZJYQDGZ-UHFFFAOYSA-N
SMILES: OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)