missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Dicoumarol

Catalog No. 287897500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
287897500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 287897500MG Supplier MilliporeSigma™ Supplier No. 287897500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable quinone reductase inhibitor with anticoagulant properties.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color White
Description 98% by titration
Target Proteins Blocks brefeldin A-dependent mono-ADP-ribosylation in vitro
CAS 66-76-2
Molecular Formula C19H12O6
Formula Weight 336.3g/mol
Solubility Aqueous alkaline solutions or organic bases
Content And Storage Protect from light
Form Solid
Quantity 500 mg