CAS RN 657-27-2
Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+%
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N Synonym: l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl PubChem CID: 69568 ChEBI: CHEBI:53633 IUPAC Name: (2S)-2,6-diaminohexanoic acid;hydrochloride SMILES: Cl.NCCCC[C@H](N)C(O)=O
TraceCERT™ L-Lysine monohydrochloride, MilliporeSigma™ Supelco™
L-Lysine monohydrochloride is generally used as a food and feed supplement.It can also find applications in medicine and pharmaceutical industries.L-Lysine monohydrochloride can be produced via sugar fermentation by modified microorganisms.
MilliporeSigma™ L-Lysine, Hydrochloride, Calbiochem™,
Format: Glass Bottle
Thermo Scientific Chemicals L(+)-Lysine monohydrochloride, 99%
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N Synonym: l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl PubChem CID: 69568 ChEBI: CHEBI:53633 SMILES: Cl.NCCCC[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N Synonym: l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl PubChem CID: 69568 ChEBI: CHEBI:53633 IUPAC Name: (2S)-2,6-diaminohexanoic acid;hydrochloride SMILES: Cl.NCCCC[C@H](N)C(O)=O
L-(+)-Lysine Monohydrochloride 98.0+%, TCI America™
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N Synonym: l-lysine hydrochloride,l-lysine monohydrochloride,lysine hydrochloride,h-lys-oh.hcl,l-lysine, monohydrochloride,lyamine,darvyl,lysion,s-2,6-diaminohexanoic acid hydrochloride,enisyl PubChem CID: 69568 ChEBI: CHEBI:53633 IUPAC Name: (2S)-2,6-diaminohexanoic acid hydrochloride SMILES: Cl.NCCCC[C@H](N)C(O)=O
Lysine Hydrochloride, Granular, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N IUPAC Name: (2S)-2,6-diaminohexanoic acid hydrochloride SMILES: Cl.NCCCC[C@H](N)C(O)=O
L-Lysine Monohydrochloride, FCC, 98.5-101.5%, Spectrum™ Chemical
CAS: 657-27-2 Molecular Formula: C6H15ClN2O2 Molecular Weight (g/mol): 182.65 MDL Number: MFCD00064564 MFCD00081870 InChI Key: BVHLGVCQOALMSV-JEDNCBNOSA-N IUPAC Name: (2S)-2,6-diaminohexanoic acid hydrochloride SMILES: Cl.NCCCC[C@H](N)C(O)=O
L-Lysine, Monochloride, Monohydrate, USP, EP, bioCERTIFIED, 98.5-101.5%, Spectrum™ Chemical
CAS: 657-27-2
Medisca L - Lysine Hydrochloride, USP
L - Lysine Hydrochloride, USP is a white, odorless powder. If using for sterile preparation, ensure further processing and testing.