CAS RN 65085-83-8
IUPAC Name:
1-[4-(benzenesulfonyl)phenyl]ethan-1-one
Synonyms:
1-[4-(benzenesulfonyl)phenyl]ethanone
Molecular Weight (g/mol):
260.31
Molecular Formula:
C14H12O3S
InChi Key:
FGFHDNIGKVTTLC-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
4-Acetyldiphenyl sulfone, 98%
4-Acetyldiphenyl sulfone, 98%
CAS: 65085-83-8 Molecular Formula: C14H12O3S Molecular Weight (g/mol): 260.307 MDL Number: MFCD00209628 InChI Key: FGFHDNIGKVTTLC-UHFFFAOYSA-N Synonym: 4-acetyldiphenyl sulfone,1-4-phenylsulfonyl phenyl ethanone,1-4-benzenesulfonyl phenyl ethanone,4-acetyl diphenyl sulfone,1-4-phenylsulfonyl phenyl ethan-1-one,pubchem15334,4-acetyldiphenylsulfone,4-phenylsulfonyl acetophenone,phenyl 4-acetylphenyl sulfone PubChem CID: 265878 IUPAC Name: 1-[4-(benzenesulfonyl)phenyl]ethanone SMILES: CC(=O)C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2