missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyldiphenyl sulfone, 98%

Catalog No. AAA1837614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1837614 25 g
AAA1837603 1 g
AAA1837606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1837614 Supplier Thermo Scientific Chemicals Supplier No. A1837614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65085-83-8 | C14H12O3S | 260.307 g/mol

4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 65085-83-8
Molecular Formula C14H12O3S
Molecular Weight (g/mol) 260.307
MDL Number MFCD00209628
InChI Key FGFHDNIGKVTTLC-UHFFFAOYSA-N
Synonym 4-acetyldiphenyl sulfone, 1-4-phenylsulfonyl phenyl ethanone, 1-4-benzenesulfonyl phenyl ethanone, 4-acetyl diphenyl sulfone, 1-4-phenylsulfonyl phenyl ethan-1-one, pubchem15334, 4-acetyldiphenylsulfone, 4-phenylsulfonyl acetophenone, phenyl 4-acetylphenyl sulfone
PubChem CID 265878
IUPAC Name 1-[4-(benzenesulfonyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 138°C to 140°C
Quantity 25 g
Formula Weight 260.31
Percent Purity 98%
Chemical Name or Material 4-Acetyldiphenyl sulfone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only