accessibility menu, dialog, popup

UserName

NH2OHOHO

CAS RN 65-49-6

IUPAC Name: 4-amino-2-hydroxybenzoic acid
Synonyms: aminosalicylic acid pas-C entepas pasem paser sanipirol-4 rezipas pas - para-aminosalicylic acid para-aminosodium salicylate alumino-4-aminosalicylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: WUBBRNOQWQTFEX-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)