accessibility menu, dialog, popup

UserName

CAS RN 6485-79-6

IUPAC Name: tris(propan-2-yl)silane
Synonyms: triisopropylsilane
Molecular Weight (g/mol): 158.36
Molecular Formula: C9H22Si
InChi Key: YDJXDYKQMRNUSA-UHFFFAOYSA-N
SMILES: CC(C)[SiH](C(C)C)C(C)C
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)