missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilane 98.0+%, TCI America™

Catalog No. T15335ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15335ML 5 mL
T153325ML 25 mL
T1533100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T15335ML Supplier TCI America Supplier No. T15335ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6485-79-6
Molecular Formula C9H22Si
Molecular Weight (g/mol) 158.36
MDL Number MFCD00009657
InChI Key YDJXDYKQMRNUSA-UHFFFAOYSA-N
Synonym triisopropylsilane, tris propan-2-yl silane, triisopropyl silane, tri propan-2-yl silicon, tri-isopropylsilyl radical, triisopropylsilyl, tri-iso-propylsilane, ambotzrl-1102, pubchem12855
PubChem CID 6327611
IUPAC Name tris(propan-2-yl)silane
SMILES CC(C)[SiH](C(C)C)C(C)C

Specifications

Color Colorless
Boiling Point 166°C
Quantity 5 mL
UN Number 1993
Formula Weight 158.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropylsilane