accessibility menu, dialog, popup

UserName

CAS RN 64697-40-1

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium chloride
Synonyms: 1-octyl-3-methylimidazolium chloride
Molecular Weight (g/mol): 230.78
Molecular Formula: C12H23ClN2
InChi Key: OXFBEEDAZHXDHB-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)