missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Chloride 98.0+%, TCI America™

Catalog No. M20625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20625G 5 g
M206225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20625G Supplier TCI America Supplier No. M20625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64697-40-1
Molecular Formula C12H23ClN2
Molecular Weight (g/mol) 230.78
MDL Number MFCD03095432
InChI Key OXFBEEDAZHXDHB-UHFFFAOYSA-M
Synonym 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
PubChem CID 2734223
ChEBI CHEBI:61342
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium chloride
SMILES [Cl-].CCCCCCCCN1C=C[N+](C)=C1

Specifications

Melting Point 12°C
Quantity 5 g
Formula Weight 230.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Chloride