accessibility menu, dialog, popup

UserName

CAS RN 646-04-8

IUPAC Name: (2E)-pent-2-ene
Synonyms: trans-2-pentene
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: QMMOXUPEWRXHJS-HWKANZROSA-N
SMILES: CC\C=C\C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)