missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentene 99.0+%, TCI America™

Catalog No. P063510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P063510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P063510ML Supplier TCI America Supplier No. P063510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646-04-8
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HWKANZROSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
IUPAC Name (E)-pent-2-ene
SMILES CCC=CC

Specifications

Color Yellow
Boiling Point 36°C
Quantity 10 mL
UN Number 3295
Formula Weight 70.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Pentene
EINECSNumber (2)-0019
TSCA Yes