accessibility menu, dialog, popup

UserName

H3CNSClOCl

CAS RN 64359-81-5

IUPAC Name: 4,5-dichloro-2-octyl-2,3-dihydro-1,2-thiazol-3-one
Synonyms: 4,5-dichloro-2-octyl-1,2-thiazol-3-one
Molecular Weight (g/mol): 282.22
Molecular Formula: C11H17Cl2NOS
InChi Key: PORQOHRXAJJKGK-UHFFFAOYSA-N
SMILES: CCCCCCCCN1SC(Cl)=C(Cl)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results