missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one 98.0+%, TCI America™

Catalog No. D415725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D415725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D415725G Supplier TCI America Supplier No. D415725G
Only null left

Chemical Identifiers

CAS 64359-81-5
Molecular Formula C11H17Cl2NOS
Molecular Weight (g/mol) 282.22
MDL Number MFCD04034673
InChI Key PORQOHRXAJJKGK-UHFFFAOYSA-N
PubChem CID 91688
ChEBI CHEBI:83518
IUPAC Name 4,5-dichloro-2-octyl-2,3-dihydro-1,2-thiazol-3-one
SMILES CCCCCCCCN1SC(Cl)=C(Cl)C1=O

Specifications

Melting Point 42°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 282.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one
EINECSNumber (5)-6165
RTECSNumber NX8157075
TSCA Yes