accessibility menu, dialog, popup

UserName

CAS RN 6422-86-2

IUPAC Name: 1,4-bis(2-ethylhexyl) benzene-1,4-dicarboxylate
Synonyms: dioctyl terephthalate diethylhexyl terephthalate
Molecular Weight (g/mol): 390.56
Molecular Formula: C24H38O4
InChi Key: RWPICVVBGZBXNA-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC
Boiling Point 
  • (3)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results