accessibility menu, dialog, popup

UserName

BrOO

CAS RN 64169-34-2

IUPAC Name: 5-bromo-1,3-dihydro-2-benzofuran-1-one
Synonyms: 5-bromo-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 213.03
Molecular Formula: C8H5BrO2
InChi Key: IUSPXLCLQIZFHL-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)OCC2=C1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)