missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromophthalide 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 165°C |
| Color | Blanc-jaune |
| Quantity | 25 g |
| Formula Weight | 213.03 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 5-Bromophthalide |
Sécurité et manipulation
| TSCA | No |