CAS RN 64039-28-7
IUPAC Name:
sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate
Synonyms:
sodium 3-acetyl-6-methyl-2,4-dioxo-3H-pyran-3-ide hydrate
Molecular Weight (g/mol):
208.15
Molecular Formula:
C8H9NaO5
InChi Key:
AFIQQQWSAQPUQO-UHFFFAOYSA-N
SMILES:
O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O
1
–
2
of
2
results
2
Sodium Dehydroacetate, 98-100.5%, Spectrum™ Chemical
CAS: 64039-28-7 Molecular Formula: C8H9NaO5 Molecular Weight (g/mol): 208.15 InChI Key: AFIQQQWSAQPUQO-UHFFFAOYSA-N IUPAC Name: sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide hydrate SMILES: O.[Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O