missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dehydroacetate Monohydrate 98.0+%, TCI America™

Catalog No. D004025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D004025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D004025G Supplier TCI America Supplier No. D004025G
Only null left

Specifications

CAS 64039-28-7
Color White-Yellow
Molecular Formula C8H8O4
Quantity 25 g
MDL Number MFCD00040583
Synonym 3-Acetyl-4-hydroxy-6-methyl-2-pyrone Sodium Salt, Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Dehydroacetic Acid Sodium Salt
Formula Weight 190.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dehydroacetate Monohydrate
EINECSNumber (5)-0665
RTECSNumber UP8400000
TSCA Yes