accessibility menu, dialog, popup

UserName

CH3OO(S)OH3COCH3OH3CNHCH3O

CAS RN 64-86-8

IUPAC Name: N-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
Synonyms: colchicine colchicine, its salts and derivatives colchicina colchicin colchicinum condylon (S)-colchicine 7alphah-colchicine
Molecular Weight (g/mol): 399.44
Molecular Formula: C22H25NO6
InChi Key: IAKHMKGGTNLKSZ-INIZCTEOSA-N
SMILES: COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (4)
  • (3)
Physical Form 
  • (2)
  • (6)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (9)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (6)
  • (1)