missing translation for 'onlineSavingsMsg'
Learn More

Colchicine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017776
Change view
Click to view available options
Quantity:
400 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017776 400 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017776 Supplier Merck Emd Millipore Supplier No. PHR1764400MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Colchicine is a drug used for the treatment of gout and certain rheumatic and nonrheumatic conditions. It is the major alkaloid in the plant Colchicum autumnale (meadow saffron). Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 64-86-8
Linear Formula C22H25 NO6
MDL Number MFCD00078484
Melting Point 150°C to 160°C
Synonym Colchicine; (S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide
Type Certified Reference Material
Quantity 400 mg
Formula Weight 399.44
Grade Certified Reference Material