missing translation for 'onlineSavingsMsg'
Learn More

Colchicine (contains 5% Ethyl Acetate at maximum) 97.0+%, TCI America™

Catalog No. C03805G
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03805G 5 g
C0380500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03805G Supplier TCI America Supplier No. C03805G

Chemical Identifiers

CAS 64-86-8
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.44
MDL Number MFCD00078484,MFCD00078484
InChI Key IAKHMKGGTNLKSZ-INIZCTEOSA-N
Synonym colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
PubChem CID 45038708
IUPAC Name N-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
SMILES COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1

Specifications

Melting Point 150°C
Color Yellow
Quantity 5 g
UN Number 1544
Formula Weight 399.44
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Colchicine (contains 5% Ethyl Acetate at maximum)
RTECSNumber GH0700000
TSCA Yes