CAS RN 64-77-7
IUPAC Name:
3-butyl-1-(4-methylbenzenesulfonyl)urea
Synonyms:
tolbutamide dolipol orinase diagnostic drabet restinon willbutamide diabetamid tolylsulfonylbutylurea orezan tolbutamida
Molecular Weight (g/mol):
270.35
Molecular Formula:
C12H18N2O3S
InChi Key:
JLRGJRBPOGGCBT-UHFFFAOYSA-N
SMILES:
CCCCNC(=O)NS(=O)(=O)C1=CC=C(C)C=C1
1
–
2
of
2
results
2
Tolbutamide, 98%
CAS: 64-77-7 Molecular Formula: C12H18N2O3S Molecular Weight (g/mol): 270.347 MDL Number: MFCD00027169 InChI Key: JLRGJRBPOGGCBT-UHFFFAOYSA-N Synonym: tolbutamide,orinase,1-butyl-3-tosylurea,tolbutamid,aglicid,arkozal,artosin,diabetol,dolipol,rastinon PubChem CID: 5505 ChEBI: CHEBI:27999 IUPAC Name: 1-butyl-3-(4-methylphenyl)sulfonylurea SMILES: CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C