missing translation for 'onlineSavingsMsg'
Learn More

Tolbutamide, 98%

Catalog No. AAB2169830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169830 250 g
AAB2169809 10 g
AAB2169818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2169830 Supplier Thermo Scientific Chemicals Supplier No. B2169830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-77-7 | C12H18N2O3S | 270.347 g/mol

Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 64-77-7
Molecular Formula C12H18N2O3S
Molecular Weight (g/mol) 270.347
MDL Number MFCD00027169
InChI Key JLRGJRBPOGGCBT-UHFFFAOYSA-N
Synonym tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon
PubChem CID 5505
ChEBI CHEBI:27999
IUPAC Name 1-butyl-3-(4-methylphenyl)sulfonylurea
SMILES CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 128°C to 130°C
Quantity 250 g
Beilstein 1984428
Merck Index 14,9507
Solubility Information Soluble in chloroform.
Formula Weight 270.35
Percent Purity 98%
Chemical Name or Material Tolbutamide
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 200-594-3
RTECSNumber YS4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only