missing translation for 'onlineSavingsMsg'
Learn More

Tolbutamide, MilliporeSigma™ Supelco™

Catalog No. 111015064
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015064 100 g
111015063 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015064 Supplier Merck Emd Millipore Supplier No. T0891100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 64-77-7
Physical Form Neat
Linear Formula C12H18N2O3 S
MDL Number MFCD00027169
Synonym 1-Butyl-3-(4-methylphenylsulfonyl)urea; N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide
Type Analytical Standard
Quantity 100 g
Formula Weight 270.35
Grade Analytical Standard