accessibility menu, dialog, popup

UserName

FFFNHN

CAS RN 63854-31-9

IUPAC Name: 1-[2-(trifluoromethyl)phenyl]piperazine
Synonyms: trifluoromethylphenylpiperazine
Molecular Weight (g/mol): 230.23
Molecular Formula: C11H13F3N2
InChi Key: VZUBMIDXJRGARE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1N1CCNCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results