missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Trifluoromethylphenyl)piperazine 98.0+%, TCI America™

Catalog No. T27395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27395G 5 g
T27391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27395G Supplier TCI America Supplier No. T27395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63854-31-9
Molecular Formula C11H13F3N2
Molecular Weight (g/mol) 230.234
MDL Number MFCD02094727
InChI Key VZUBMIDXJRGARE-UHFFFAOYSA-N
Synonym 2-(1-Piperazinyl)benzotrifluoride
PubChem CID 2777670
IUPAC Name 1-[2-(trifluoromethyl)phenyl]piperazine
SMILES C1CN(CCN1)C2=CC=CC=C2C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 230.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Trifluoromethylphenyl)piperazine