accessibility menu, dialog, popup

UserName

CAS RN 63799-11-1

H3C(S)H3CN

CAS RN 63799-11-1

IUPAC Name: 4'-[(2S)-2-methylbutyl]-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-[(2S)-2-methylbutyl]-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 249.36
Molecular Formula: C18H19N
InChi Key: DNJQGRFZQMOYGM-AWEZNQCLSA-N
SMILES: CC[C@H](C)CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)