accessibility menu, dialog, popup

UserName

CAS RN 637-55-8

IUPAC Name: phenyl octadecanoate
Synonyms: octadecanoic acid, phenyl ester
Molecular Weight (g/mol): 360.58
Molecular Formula: C24H40O2
InChi Key: NUMNZKICGJJSHN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results