missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Stearate 98.0+%, TCI America™

Catalog No. O00075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00075G 5 g
O000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00075G Supplier TCI America Supplier No. O00075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-55-8
Molecular Formula C24H40O2
Molecular Weight (g/mol) 360.58
MDL Number MFCD00026663
InChI Key NUMNZKICGJJSHN-UHFFFAOYSA-N
Synonym phenyl stearate, octadecanoic acid, phenyl ester, stearic acid phenyl ester, stearic acid, phenyl ester, phenylstearate, acmc-209nhu, octadecanoicacid,phenylester, stearic acid, phenyl ester 8ci
PubChem CID 69477
IUPAC Name phenyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC1=CC=CC=C1

Specifications

Melting Point 53°C
Color White
Boiling Point 267°C
Quantity 5 g
Formula Weight 360.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Stearate