accessibility menu, dialog, popup

UserName

CAS RN 637-54-7

IUPAC Name: N-phenyloctadecanamide
Synonyms: N-phenyloctadecanamide
Molecular Weight (g/mol): 359.6
Molecular Formula: C24H41NO
InChi Key: ZOLJFBQEKSZVCB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results