accessibility menu, dialog, popup

UserName

CH3OOCH2

CAS RN 63600-35-1

IUPAC Name: 2-ethenylphenyl acetate
Synonyms: 2-ethenylphenyl acetate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: WRPYDXWBHXAKPT-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results