accessibility menu, dialog, popup

UserName

CAS RN 636-72-6

IUPAC Name: (thiophen-2-yl)methanol
Synonyms: 2-hydroxymethylthiophene
Molecular Weight (g/mol): 114.16
Molecular Formula: C5H6OS
InChi Key: ZPHGMBGIFODUMF-UHFFFAOYSA-N
SMILES: OCC1=CC=CS1
Boiling Point 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (6)