missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethanol, 97%

Catalog No. AAA1576822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1576822 100 g
AAA1576814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1576822 Supplier Thermo Scientific Chemicals Supplier No. A1576822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 636-72-6 | C5H6OS | 114.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-72-6
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00005454
InChI Key ZPHGMBGIFODUMF-UHFFFAOYSA-N
Synonym 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
PubChem CID 69467
IUPAC Name thiophen-2-ylmethanol
SMILES OCC1=CC=CS1

Specifications

Melting Point 0°C
Density 1.206
Boiling Point 206°C to 207°C
Flash Point >110°C (230°F)
Refractive Index 1.565
Quantity 100 g
Beilstein 383595
Sensitivity Air Sensitive
Formula Weight 114.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethanol
Show More Show Less
Hazard Category H302-H312-H315-H319-H332-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-264-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only