accessibility menu, dialog, popup

UserName

CAS RN 635-27-8

ClN

CAS RN 635-27-8

IUPAC Name: 5-chloroquinoline
Synonyms: 5-chloroquinoline
Molecular Weight (g/mol): 163.6
Molecular Formula: C9H6ClN
InChi Key: HJSRGOVAIOPERP-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=NC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results