missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroquinoline 98.0+%, TCI America™

Catalog No. C29975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29975G 5 g
C29971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29975G Supplier TCI America Supplier No. C29975G
Only null left

Chemical Identifiers

CAS 635-27-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00239413
InChI Key HJSRGOVAIOPERP-UHFFFAOYSA-N
PubChem CID 69458
IUPAC Name 5-chloroquinoline
SMILES C1=CC2=C(C=CC=N2)C(=C1)Cl

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 263°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroquinoline
TSCA No
Recommended Storage Refrigerator