accessibility menu, dialog, popup

UserName

CAS RN 633-65-8

IUPAC Name: 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium chloride
Synonyms: berberine chloride
Molecular Weight (g/mol): 371.82
Molecular Formula: C20H18ClNO4
InChi Key: VKJGBAJNNALVAV-UHFFFAOYSA-M
SMILES: [Cl-].COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC
Form 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results