accessibility menu, dialog, popup

UserName

CAS RN 6305-95-9

IUPAC Name: 1-(2-chlorophenyl)propan-2-one
Synonyms: 1-(2-chlorophenyl)propan-2-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: LWGNDIMNCPMZOF-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results