accessibility menu, dialog, popup

UserName

CAS RN 630-76-2

IUPAC Name: (triphenylmethyl)benzene
Synonyms: tetraphenylmethane
Molecular Weight (g/mol): 320.44
Molecular Formula: C25H20
InChi Key: PEQHIRFAKIASBK-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)